Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-(methylsulfonyl)benzoic acid, 96%

Click to view available options
Quantity:
1g
2.5g
53250-83-2
C8H7ClO4S
234.65
MFCD00216496
CTTWSFIIFMWHLQ-UHFFFAOYSA-N
2-chloro-4-methylsulfonyl benzoic acid,2-chloro-4-methylsulphonylbenzoic acid,2-chloro-4-methanesulfonylbenzoic acid,benzoic acid, 2-chloro-4-methylsulfonyl,2-chloro-4-methanesulfonyl-benzoic acid,2-chloro-4-methylsulfonyl benzoic acid dry basis,maybridge1_004133,cmba,pubchem20281,acmc-1aupa
735863
2-chloro-4-methylsulfonylbenzoic acid
CS(=O)(=O)C1=CC(=C(C=C1)C(=O)O)Cl
53250-83-2
C8H7ClO4S
234.65
MFCD00216496
CTTWSFIIFMWHLQ-UHFFFAOYSA-N
2-chloro-4-methylsulfonyl benzoic acid,2-chloro-4-methylsulphonylbenzoic acid,2-chloro-4-methanesulfonylbenzoic acid,benzoic acid, 2-chloro-4-methylsulfonyl,2-chloro-4-methanesulfonyl-benzoic acid,2-chloro-4-methylsulfonyl benzoic acid dry basis,maybridge1_004133,cmba,pubchem20281,acmc-1aupa
735863
2-chloro-4-methylsulfonylbenzoic acid
CS(=O)(=O)C1=CC(=C(C=C1)C(=O)O)Cl
Melting Point 191°C to 196°C
Quantity 2.5g
Formula Weight 234.66
Percent Purity 96%
Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280i-P305+P351+P338
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger