Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-nitroaniline, 98+%

Click to view available options
Quantity:
250g
1000g
5000g
121-87-9
C6H5ClN2O2
172.568
MFCD00007665
LOCWBQIWHWIRGN-UHFFFAOYSA-N
ocpna,2-chloro-4-nitrobenzenamine,o-chloro-p-nitroaniline,4-nitro-2-chloroaniline,benzenamine, 2-chloro-4-nitro,1-amino-2-chloro-4-nitrobenzene,aniline, 2-chloro-4-nitro,unii-xsa3zx337b,2-chloro-4-nitro-phenylamine,ortho-chloro-para-nitroaniline
8492
2-chloro-4-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])Cl)N
121-87-9
C6H5ClN2O2
172.568
MFCD00007665
LOCWBQIWHWIRGN-UHFFFAOYSA-N
ocpna,2-chloro-4-nitrobenzenamine,o-chloro-p-nitroaniline,4-nitro-2-chloroaniline,benzenamine, 2-chloro-4-nitro,1-amino-2-chloro-4-nitrobenzene,aniline, 2-chloro-4-nitro,unii-xsa3zx337b,2-chloro-4-nitro-phenylamine,ortho-chloro-para-nitroaniline
8492
2-chloro-4-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])Cl)N
Boiling Point 320°C (decomposition)
Flash Point 193°C (379°F)
Quantity 250g
Melting Point 106°C to 109°C
Odor Weak, Characteristic
UN Number UN2237
Beilstein 638657
Formula Weight 172.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-4-nitroaniline

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES

EINECSNumber : 204-502-2

RTECSNumber : BX1400000

TSCA : Yes

Recommended Storage : Ambient temperatures