Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-nitrobenzamide, 98%

Click to view available options
Quantity:
5g
25g
3011-89-0
C7H5ClN2O3
200.578
MFCD00017119
GFGSZUNNBQXGMK-UHFFFAOYSA-N
aklomide,aklomix,clomide,alkomide,benzamide, 2-chloro-4-nitro,component of aklomix,component of novastat-w,unii-b0e341ra20,2-chloro-4-nitro benzamide,dsstox_cid_21405
2075
2-chloro-4-nitrobenzamide
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N
3011-89-0
C7H5ClN2O3
200.578
MFCD00017119
GFGSZUNNBQXGMK-UHFFFAOYSA-N
aklomide,aklomix,clomide,alkomide,benzamide, 2-chloro-4-nitro,component of aklomix,component of novastat-w,unii-b0e341ra20,2-chloro-4-nitro benzamide,dsstox_cid_21405
2075
2-chloro-4-nitrobenzamide
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N
Quantity 25g
Melting Point 168°C to 172°C
Beilstein 1966537
Formula Weight 200.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrobenzamide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 221-143-7

TSCA : No

Recommended Storage : Ambient temperatures