Alfa Aesar™ 2-Chloro-4-nitrobenzoic acid, 98%

Manufacturer: Alfa Aesar™ A1160236

Quantity 500g

 View more versions of this product

Catalog No. AAA1160236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99-60-5
    Molecular Formula C7H4ClNO4
    Formula Weight 201.57
    MDL Number MFCD00007209
    Beilstein 1912836
    Chemical Name or Material 2-Chloro-4-nitrobenzoic acid

    Specifications

    Formula Weight 201.57
    Health Hazard 3 GHS P Statement
    P280a-P305+P351+P338-P337+P313
    Wear protective gloves and eye/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Odor Odorless
    Health Hazard 1 Danger
    TSCA TSCA
    MDL Number MFCD00007209
    CAS 99-60-5
    RTECS Number DG5425000
    Melting Point 140° to 144°C
    Quantity 500g
    Molecular Formula C7H4ClNO4
    Beilstein 1912836
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    Chemical Name or Material 2-Chloro-4-nitrobenzoic acid
    Assay 98%

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P280a-P305+P351+P338-P337+P313
    Wear protective gloves and eye/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit