Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-nitrobenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
28163-00-0
C7H3ClN2O2
182.563
MFCD00052203
ZIGQFRQZYVQQDR-UHFFFAOYSA-N
2-chloro-4-nitro-benzonitrile,benzonitrile, 2-chloro-4-nitro,2-chloro-4-nitrobenzenecarbonitrile,pubchem18759,acmc-1ceg9,2-chloro-4-nitrobenzonitril,chloronitrobenzenecarbonitrile,3-chloro-4-cyanonitrobenzene,ksc497k1r
141401
2-chloro-4-nitrobenzonitrile
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C#N
28163-00-0
C7H3ClN2O2
182.563
MFCD00052203
ZIGQFRQZYVQQDR-UHFFFAOYSA-N
2-chloro-4-nitro-benzonitrile,benzonitrile, 2-chloro-4-nitro,2-chloro-4-nitrobenzenecarbonitrile,pubchem18759,acmc-1ceg9,2-chloro-4-nitrobenzonitril,chloronitrobenzenecarbonitrile,3-chloro-4-cyanonitrobenzene,ksc497k1r
141401
2-chloro-4-nitrobenzonitrile
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C#N
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Beige
Melting Point 78°C to 82°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(NO2)CN
Formula Weight 182.57
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloro-4-nitrobenzonitrile
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning