Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-nitrobenzotrifluoride, 98%

Click to view available options
Quantity:
1g
5g
151504-80-2
C7H3ClF3NO2
225.551
MFCD00171532
MOTWHSMEZCFDOD-UHFFFAOYSA-N
2-chloro-4-nitrobenzotrifluoride,2-chloro-4-nitro-1-trifluoromethyl benzene,benzene, 2-chloro-4-nitro-1-trifluoromethyl,pubchem4852,intermediates-zcf02158,3-chloro-4-trifluoromethyl-nitrobenzene,3-chloro-4-trifluoromethyl-1-nitrobenzene,2-chloranyl-4-nitro-1-trifluoromethyl benzene
2778985
2-chloro-4-nitro-1-(trifluoromethyl)benzene
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(F)(F)F
151504-80-2
C7H3ClF3NO2
225.551
MFCD00171532
MOTWHSMEZCFDOD-UHFFFAOYSA-N
2-chloro-4-nitrobenzotrifluoride,2-chloro-4-nitro-1-trifluoromethyl benzene,benzene, 2-chloro-4-nitro-1-trifluoromethyl,pubchem4852,intermediates-zcf02158,3-chloro-4-trifluoromethyl-nitrobenzene,3-chloro-4-trifluoromethyl-1-nitrobenzene,2-chloranyl-4-nitro-1-trifluoromethyl benzene
2778985
2-chloro-4-nitro-1-(trifluoromethyl)benzene
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(F)(F)F
Color Colorless to Yellow
Quantity 5g
Beilstein 3311567
Formula Weight 225.55
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrobenzotrifluoride

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning