Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-nitropyridine, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
23056-36-2
C5H3ClN2O2
158.54
LIEPVGBDUYKPLC-UHFFFAOYSA-N
pyridine, 2-chloro-4-nitro,2-chloro-4-nitro-pyridine,2-chlor-4-nitropyridin,pubchem9220,2-chloro-4nitropyridine,2chloro-4-nitropyridine,acmc-1ccv7,2-chloro-4-nitro pyridine,ksc201q1f,2-chloro-4-nitropyridine
735152
2-chloro-4-nitropyridine
C1=CN=C(C=C1[N+](=O)[O-])Cl
23056-36-2
C5H3ClN2O2
158.54
LIEPVGBDUYKPLC-UHFFFAOYSA-N
pyridine, 2-chloro-4-nitro,2-chloro-4-nitro-pyridine,2-chlor-4-nitropyridin,pubchem9220,2-chloro-4nitropyridine,2chloro-4-nitropyridine,acmc-1ccv7,2-chloro-4-nitro pyridine,ksc201q1f,2-chloro-4-nitropyridine
735152
2-chloro-4-nitropyridine
C1=CN=C(C=C1[N+](=O)[O-])Cl
Packaging Glass Bottle
Quantity 1g
Melting Point 52.0°C to 56.0°C
Assay Percent Range 97.5% min. (GC)
Formula Weight 158.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitropyridine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning