Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-nitropyridine N-oxide, 97%

Click to view available options
Quantity:
10g
50g
14432-16-7
C5H3ClN2O3
174.54
MFCD00955617
YSTCMHHKDOVZDA-UHFFFAOYSA-N
2-chloro-4-nitropyridine-n-oxide,2-chloro-4-nitropyridine 1-oxide,2-chloro-4-nitropyridine n-oxide,2-chloro-4-nitropyridine1-oxide,2-chloro-4-nitropyridin-1-ium-1-olate,2-chloro-4-nitropyridin-1-ol,2-chlor-4-nitropyridin-1-oxid,pubchem1191,acmc-1cisj,pycl12
84433
2-chloro-4-nitro-1-oxidopyridin-1-ium
C1=C[N+](=C(C=C1[N+](=O)[O-])Cl)[O-]
14432-16-7
C5H3ClN2O3
174.54
MFCD00955617
YSTCMHHKDOVZDA-UHFFFAOYSA-N
2-chloro-4-nitropyridine-n-oxide,2-chloro-4-nitropyridine 1-oxide,2-chloro-4-nitropyridine n-oxide,2-chloro-4-nitropyridine1-oxide,2-chloro-4-nitropyridin-1-ium-1-olate,2-chloro-4-nitropyridin-1-ol,2-chlor-4-nitropyridin-1-oxid,pubchem1191,acmc-1cisj,pycl12
84433
2-chloro-4-nitro-1-oxidopyridin-1-ium
C1=C[N+](=C(C=C1[N+](=O)[O-])Cl)[O-]
Melting Point 152°C to 155°C
Odor Odorless
Quantity 50g
Beilstein 145697
Formula Weight 174.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-nitropyridine N-oxide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning