Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-nitrotoluene, 98%

Click to view available options
Quantity:
25g
100g
500g
121-86-8
C7H6ClNO2
171.58
MFCD00007210
LLYXJBROWQDVMI-UHFFFAOYSA-N
2-chloro-4-nitrotoluene,benzene, 2-chloro-1-methyl-4-nitro,o-chloro-p-nitrotoluol,toluene, 2-chloro-4-nitro,3-chloro-4-methylnitrobenzene,2-chlor-4-nitrotoluen,unii-9t26lnr6t8,2-chloro-4-nitrotoluen czech,2,4-chloronitrotoluene,2-chloro-4-nitro-1-methylbenzene
8491
2-chloro-1-methyl-4-nitrobenzene
CC1=C(C=C(C=C1)[N+](=O)[O-])Cl
121-86-8
C7H6ClNO2
171.58
MFCD00007210
LLYXJBROWQDVMI-UHFFFAOYSA-N
2-chloro-4-nitrotoluene,benzene, 2-chloro-1-methyl-4-nitro,o-chloro-p-nitrotoluol,toluene, 2-chloro-4-nitro,3-chloro-4-methylnitrobenzene,2-chlor-4-nitrotoluen,unii-9t26lnr6t8,2-chloro-4-nitrotoluen czech,2,4-chloronitrotoluene,2-chloro-4-nitro-1-methylbenzene
8491
2-chloro-1-methyl-4-nitrobenzene
CC1=C(C=C(C=C1)[N+](=O)[O-])Cl
Quantity 500g
Melting Point 62°C to 66°C
Boiling Point 260°C
UN Number UN3457
Beilstein 1817924
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrotoluene

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID

RTECSNumber : XS9100000

TSCA : TSCA