Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
3678-62-4
C6H6ClN
127.57
MFCD00023418
MZVSTDHRRYQFGI-UHFFFAOYSA-N
2-chloro-4-picoline,pyridine, 2-chloro-4-methyl,2-chloro-4-methyl-pyridine,2-chloro-4-methyl pyridine,2-chloro-gamma-picoline,4-picoline, 2-chloro,zlchem 438,pubchem1192,acmc-209iop
77248
2-chloro-4-methylpyridine
CC1=CC(=NC=C1)Cl
3678-62-4
C6H6ClN
127.57
MFCD00023418
MZVSTDHRRYQFGI-UHFFFAOYSA-N
2-chloro-4-picoline,pyridine, 2-chloro-4-methyl,2-chloro-4-methyl-pyridine,2-chloro-4-methyl pyridine,2-chloro-gamma-picoline,4-picoline, 2-chloro,zlchem 438,pubchem1192,acmc-209iop
77248
2-chloro-4-methylpyridine
CC1=CC(=NC=C1)Cl
Flash Point 89°C
Boiling Point 194°C to 195°C
Specific Gravity 1.151
Density 1.1510g/mL
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 25mL
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 20,241
Formula Weight 127.57
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-picoline
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 222-951-2