Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-(trifluoromethoxy)aniline, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
69695-61-0
C7H5ClF3NO
211.57
MFCD01631704
YMLWONHDNGICOH-UHFFFAOYSA-N
2-chloro-4-trifluoromethoxy aniline,benzenamine, 2-chloro-4-trifluoromethoxy,2-chloro-4-trifluoromethoxy phenylamine,benzenamine,2-chloro-4-trifluoromethoxy,2-chloro-4-trifluoromethoxy-phenylamine,pubchem2941,acmc-209wey,ksc630s5l,2-chloro4-trifluoromethoxyaniline,4-trifluoromethoxy-2-chloroaniline
2734084
2-chloro-4-(trifluoromethoxy)aniline
C1=CC(=C(C=C1OC(F)(F)F)Cl)N
69695-61-0
C7H5ClF3NO
211.57
MFCD01631704
YMLWONHDNGICOH-UHFFFAOYSA-N
2-chloro-4-trifluoromethoxy aniline,benzenamine, 2-chloro-4-trifluoromethoxy,2-chloro-4-trifluoromethoxy phenylamine,benzenamine,2-chloro-4-trifluoromethoxy,2-chloro-4-trifluoromethoxy-phenylamine,pubchem2941,acmc-209wey,ksc630s5l,2-chloro4-trifluoromethoxyaniline,4-trifluoromethoxy-2-chloroaniline
2734084
2-chloro-4-(trifluoromethoxy)aniline
C1=CC(=C(C=C1OC(F)(F)F)Cl)N
Refractive Index 1.4790 to 1.4830
Packaging Glass bottle
Flash Point 98°C
Quantity 5g
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Density 98g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 211.57
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning