Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Chloro-4'-(trifluoromethyl)acetanilide, 96%

Click to view available options
Quantity:
1g
5g
247170-19-0
C9H7ClF3NO
237.606
MFCD01313231
QRMZQPODCUCPQD-UHFFFAOYSA-N
n-2-chloro-4-trifluoromethyl phenyl acetamide,2'-chloro-4'-trifluoromethyl acetanilide,2-chloro-4-trifluoromethyl acetamilide,2-chloro-4-trifluoromethyl acetanilide,maybridge1_007797,n-2-chloranyl-4-trifluoromethyl phenyl ethanamide
2736671
N-[2-chloro-4-(trifluoromethyl)phenyl]acetamide
CC(=O)NC1=C(C=C(C=C1)C(F)(F)F)Cl
247170-19-0
C9H7ClF3NO
237.606
MFCD01313231
QRMZQPODCUCPQD-UHFFFAOYSA-N
n-2-chloro-4-trifluoromethyl phenyl acetamide,2'-chloro-4'-trifluoromethyl acetanilide,2-chloro-4-trifluoromethyl acetamilide,2-chloro-4-trifluoromethyl acetanilide,maybridge1_007797,n-2-chloranyl-4-trifluoromethyl phenyl ethanamide
2736671
N-[2-chloro-4-(trifluoromethyl)phenyl]acetamide
CC(=O)NC1=C(C=C(C=C1)C(F)(F)F)Cl
Quantity 5g
Melting Point 134°C to 138°C
Formula Weight 237.61
Percent Purity 96%
Chemical Name or Material 2'-Chloro-4'-(trifluoromethyl)acetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning