Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 97%, AcroSeal™, Thermo Scientific™

Catalog Number ac427840500
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
2428-06-0
C5H10ClO2P
168.56
MFCD03093988
RAOZVFGTRXLBTA-UHFFFAOYSA-N
2-chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 1,3,2-dioxaphosphorinane, 2-chloro-5,5-dimethyl, phosphorochloridous acid, cyclic 2,2-dimethyltrimethylene ester, 2-chloro-5,5-dimethyl 1,3,2 dioxaphosphinane, 5,5-dimethyl-2-chloro-1,3,2-dioxaphosphorinane, 1,3,2-dioxaphosphorinane,2-chloro-5,5-dimethyl
75508
2-chloro-5,5-dimethyl-1,3,2-dioxaphosphinane
CC1(C)COP(Cl)OC1
2428-06-0
C5H10ClO2P
168.56
MFCD03093988
RAOZVFGTRXLBTA-UHFFFAOYSA-N
2-chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 1,3,2-dioxaphosphorinane, 2-chloro-5,5-dimethyl, phosphorochloridous acid, cyclic 2,2-dimethyltrimethylene ester, 2-chloro-5,5-dimethyl 1,3,2 dioxaphosphinane, 5,5-dimethyl-2-chloro-1,3,2-dioxaphosphorinane, 1,3,2-dioxaphosphorinane,2-chloro-5,5-dimethyl
75508
2-chloro-5,5-dimethyl-1,3,2-dioxaphosphinane
CC1(C)COP(Cl)OC1

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2150g/mL
Boiling Point 46.0°C to 47.0°C (1.6 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
Quantity 50 mL
Specific Gravity 1.215
Formula Weight 168.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-380-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit