Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-chloromethylpyridine, 90%, tech., ACROS Organics™

Click to view available options
Quantity:
5g
70258-18-3
C6H5Cl2N
162.013
MFCD00125366
SKCNYHLTRZIINA-UHFFFAOYSA-N
2-chloro-5-chloromethyl pyridine,pyridine, 2-chloro-5-chloromethyl,2-chloro-5-chloromethyl-pyridine,pubchem9210,acmc-1bl9y,ksc377c3b,2-chloro-5chloromethylpyridine,2-chloro-5-chloromethypyridine,5-chloromethyl-2-chloropyridine,6-chloro-3-chloromethylpyridine
155479
2-chloro-5-(chloromethyl)pyridine
C1=CC(=NC=C1CCl)Cl
70258-18-3
C6H5Cl2N
162.013
MFCD00125366
SKCNYHLTRZIINA-UHFFFAOYSA-N
2-chloro-5-chloromethyl pyridine,pyridine, 2-chloro-5-chloromethyl,2-chloro-5-chloromethyl-pyridine,pubchem9210,acmc-1bl9y,ksc377c3b,2-chloro-5chloromethylpyridine,2-chloro-5-chloromethypyridine,5-chloromethyl-2-chloropyridine,6-chloro-3-chloromethylpyridine
155479
2-chloro-5-(chloromethyl)pyridine
C1=CC(=NC=C1CCl)Cl
Quantity 5g
Infrared Spectrum Authentic
Color Beige
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Formula Weight 162.02
Physical Form Moist Crystals
Percent Purity ≥88%
Grade Technical
Chemical Name or Material 2-Chloro-5-chloromethylpyridine, 90%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA