Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-fluorobenzyl alcohol, 97+%

Click to view available options
Quantity:
1g
5g
261762-59-8
C7H6ClFO
160.572
MFCD01631418
VTZKFYRHXVQMQL-UHFFFAOYSA-N
2-chloro-5-fluorobenzyl alcohol,2-chloro-5-fluorophenyl methanol,2-chloro-5-fluorobenzylalcohol,benzenemethanol,2-chloro-5-fluoro,2-chloro-5-fluoro-phenyl-methanol,2-chloro-5-fluorophenyl methan-1-ol,acmc-1cguv,ksc557e5t,2-chloro-5-fluorobenzenemethanol,benzenemethanol, 2-chloro-5-fluoro
2773605
(2-chloro-5-fluorophenyl)methanol
C1=CC(=C(C=C1F)CO)Cl
261762-59-8
C7H6ClFO
160.572
MFCD01631418
VTZKFYRHXVQMQL-UHFFFAOYSA-N
2-chloro-5-fluorobenzyl alcohol,2-chloro-5-fluorophenyl methanol,2-chloro-5-fluorobenzylalcohol,benzenemethanol,2-chloro-5-fluoro,2-chloro-5-fluoro-phenyl-methanol,2-chloro-5-fluorophenyl methan-1-ol,acmc-1cguv,ksc557e5t,2-chloro-5-fluorobenzenemethanol,benzenemethanol, 2-chloro-5-fluoro
2773605
(2-chloro-5-fluorophenyl)methanol
C1=CC(=C(C=C1F)CO)Cl
Quantity 1g
Melting Point 68°C to 72°C
Formula Weight 160.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-5-fluorobenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning