Alfa Aesar™ 2-Chloro-5-fluoropyridine, 95%

Manufacturer: Alfa Aesar™ H2746206

Quantity 5g

 View more versions of this product

Catalog No. AAH2746206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31301-51-6
    Molecular Formula C5H3ClFN
    Formula Weight 131.54
    MDL Number MFCD03095332
    UN Number UN2929
    Chemical Name or Material 2-Chloro-5-fluoropyridine

    Specifications

    Formula Weight 131.54
    Molecular Formula C5H3ClFN
    MDL Number MFCD03095332
    CAS 31301-51-6
    UN Number UN2929
    Health Hazard 1 Danger
    Flash Point 42°C (108°F)
    Boiling Point 74°C (38mmHg)
    Melting Point 19°C
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H301-H310-H330-H318-H226-H315-H335
    Toxic if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
    Chemical Name or Material 2-Chloro-5-fluoropyridine
    Assay 95%

    GHS H Statement
    H301-H310-H330-H318-H226-H315-H335
    Toxic if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit