Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-5-fluoropyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
62802-42-0
C4H2ClFN2
132.53
MFCD03788197
AGYUQBNABXVWMS-UHFFFAOYSA-N
2-chloro-5-fluoro-pyrimidine,5-fluoro-2-chloropyrimidine,pyrimidine, 2-chloro-5-fluoro,2-chlor-5-fluorpyrimidin,2-chloro-5-fluoro pyrimidine,pubchem4684,acmc-1b6zl,ksc352s4h,#,2-chloro-5-fluoro-1,3-diazine
583466
2-chloro-5-fluoropyrimidine
C1=C(C=NC(=N1)Cl)F
62802-42-0
C4H2ClFN2
132.53
MFCD03788197
AGYUQBNABXVWMS-UHFFFAOYSA-N
2-chloro-5-fluoro-pyrimidine,5-fluoro-2-chloropyrimidine,pyrimidine, 2-chloro-5-fluoro,2-chlor-5-fluorpyrimidin,2-chloro-5-fluoro pyrimidine,pubchem4684,acmc-1b6zl,ksc352s4h,#,2-chloro-5-fluoro-1,3-diazine
583466
2-chloro-5-fluoropyrimidine
C1=C(C=NC(=N1)Cl)F
Boiling Point 149°C to 162°C (1013.0 hPa)
Color Colorless to Yellow
Quantity 250mg
Flash Point 65°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 132.53
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-chloro-5-fluoropyrimidine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a

GHS Signal Word: Danger