Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
887707-25-7
C6H2ClF3IN
307.439
MFCD12024302
KUASPTUGCBGXAE-UHFFFAOYSA-N
2-chloro-5-iodo-3-trifluoromethyl pyridine,2-chloro-3-trifluoromethyl-5-iodo-pyridine,2-chloro-3-trifluoromethyl-5-iodopyridine,2-choro-5-iodo-3-trifluoromethylpyridine,2-chloro-3-trifluoromethyl-5-iodopyridine,2-chloro-3-trifluoromethyl-5-iodo pyridine,2-chloro-5-iodo-3-trifluoromethyl pyridine
11500513
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
C1=C(C(=NC=C1I)Cl)C(F)(F)F
887707-25-7
C6H2ClF3IN
307.439
MFCD12024302
KUASPTUGCBGXAE-UHFFFAOYSA-N
2-chloro-5-iodo-3-trifluoromethyl pyridine,2-chloro-3-trifluoromethyl-5-iodo-pyridine,2-chloro-3-trifluoromethyl-5-iodopyridine,2-choro-5-iodo-3-trifluoromethylpyridine,2-chloro-3-trifluoromethyl-5-iodopyridine,2-chloro-3-trifluoromethyl-5-iodo pyridine,2-chloro-5-iodo-3-trifluoromethyl pyridine
11500513
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
C1=C(C(=NC=C1I)Cl)C(F)(F)F
Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
Quantity 250mg
Melting Point 43°C
Sensitivity Light Sensitive
Beilstein 10249610
Formula Weight 307.44
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning