Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Chloro-5-iodopyridine, 98%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Sensitivity | Light sensitive |
Melting Point | 95°C to 98°C |
Quantity | 1g |
Beilstein | 108889 |
Formula Weight | 239.44 |
Percent Purity | 98% |
Chemical Name or Material | 2-Chloro-5-iodopyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures