Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-iodopyridine, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
69045-79-0
C5H3ClIN
239.44
QWLGCWXSNYKKDO-UHFFFAOYSA-N
2-chloro-5-indopyridine,2-chloro-5-iodo-pyridine,pyridine, 2-chloro-5-iodo,2-chloro-5-iodo pyridine,zlchem 492,pubchem2078,acmc-1bafa,2-chloro-5 iodopyridine,2-chloro-5-iodopyridine,5-iodo-2-chloro-pyridine
4397176
2-chloro-5-iodopyridine
C1=CC(=NC=C1I)Cl
69045-79-0
C5H3ClIN
239.44
QWLGCWXSNYKKDO-UHFFFAOYSA-N
2-chloro-5-indopyridine,2-chloro-5-iodo-pyridine,pyridine, 2-chloro-5-iodo,2-chloro-5-iodo pyridine,zlchem 492,pubchem2078,acmc-1bafa,2-chloro-5 iodopyridine,2-chloro-5-iodopyridine,5-iodo-2-chloro-pyridine
4397176
2-chloro-5-iodopyridine
C1=CC(=NC=C1I)Cl
Melting Point 96.0°C to 99.0°C
Quantity 25g
Infrared Spectrum Authentic
Color Beige-Yellow to White
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 239.44
Physical Form Crystalline Powder or Flakes
Percent Purity 98+%
Chemical Name or Material 2-Chloro-5-iodopyridine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning