Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-iodopyrimidine, 95%, Acros Organics

Click to view available options
Quantity:
250mg
1g
32779-38-7
C4H2ClIN2
240.43
WSZRCNZXKKTLQE-UHFFFAOYSA-N
pyrimidine, 2-chloro-5-iodo,2-chloro-5-iodo-pyrimidine,pubchem21488,acmc-209hvo,2-cloro-5-iodopyrimidine,5-iodo-2-chloropyrimidine,ksc569e7p,pyrimidine,2-chloro-5-iodo
1714223
2-chloro-5-iodopyrimidine
C1=C(C=NC(=N1)Cl)I
32779-38-7
C4H2ClIN2
240.43
WSZRCNZXKKTLQE-UHFFFAOYSA-N
pyrimidine, 2-chloro-5-iodo,2-chloro-5-iodo-pyrimidine,pubchem21488,acmc-209hvo,2-cloro-5-iodopyrimidine,5-iodo-2-chloropyrimidine,ksc569e7p,pyrimidine,2-chloro-5-iodo
1714223
2-chloro-5-iodopyrimidine
C1=C(C=NC(=N1)Cl)I
Melting Point 135°C to 136°C
Quantity 250mg
Assay Percent Range 94% min. (GC)
Formula Weight 240.43
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-iodopyrimidine

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning