Alfa Aesar™ 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid, 98%

Manufacturer: Alfa Aesar™ H52999MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52999MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 913835-92-4
    Molecular Formula C8H8BClO4
    Formula Weight 214.41
    MDL Number MFCD08689528
    Synonym Methyl 3-borono-4-chlorobenzoate; 2-Chloro-5-(methoxycarbonyl)phenylboronic acid
    Chemical Name or Material 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid

    Specifications

    Formula Weight 214.41
    MDL Number MFCD08689528
    Melting Point 230° to 232°C
    Quantity 250mg
    Molecular Formula C8H8BClO4
    CAS 913835-92-4
    Synonym Methyl 3-borono-4-chlorobenzoate; 2-Chloro-5-(methoxycarbonyl)phenylboronic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid
    Assay 98%

    Description

    Applications
    Boronic acids are often used in metal-catalyzed cross-coupling for C-C, C-N bond formation, and C-H functionalization. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates. Other known chemistry may includes 1,4-addition to activated alkenes, 1,2-addition to carbonyls and imines, addition to anhydrides, 1,4-addition to a,ß-unsaturated ketones(4), and 1,4-addition to esters and amides.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit