Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-5-methylbenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
6342-60-5
C8H7ClO2
170.59
LEBWXJZAWTVKFL-UHFFFAOYSA-N
2-chloro-5-methylbenzole acid,6-chloro-m-toluic acid,benzoic acid, 2-chloro-5-methyl,pubchem12501,3-carboxy-4-chlorotoluene,acmc-1b72o,ksc497q1d,2-chloro-5-methylbenozic acid,2-chloro-5-methyl-benzoic acid,lebwxjzawtvkfl-uhfffaoysa
240430
2-chloro-5-methylbenzoic acid
CC1=CC(=C(C=C1)Cl)C(=O)O
6342-60-5
C8H7ClO2
170.59
LEBWXJZAWTVKFL-UHFFFAOYSA-N
2-chloro-5-methylbenzole acid,6-chloro-m-toluic acid,benzoic acid, 2-chloro-5-methyl,pubchem12501,3-carboxy-4-chlorotoluene,acmc-1b72o,ksc497q1d,2-chloro-5-methylbenozic acid,2-chloro-5-methyl-benzoic acid,lebwxjzawtvkfl-uhfffaoysa
240430
2-chloro-5-methylbenzoic acid
CC1=CC(=C(C=C1)Cl)C(=O)O
Packaging Glass bottle
Quantity 25g
Color White to Beige
Infrared Spectrum Authentic
Melting Point 148.0°C to 151.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CH3C6H3(Cl)COOH
Formula Weight 170.59
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-chloro-5-methylbenzoic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning