Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-methylpyridine-3-boronic acid, 98%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
913835-86-6
C6H7BClNO2
171.39
ZMHWEBDZKPQNHG-UHFFFAOYSA-N
2-chloro-5-methylpyridine-3-boronic acid,2-chloro-5-methylpyridin-3-yl boronic acid,2-chloro-5-picoline-3-boronic acid,2-chloro-5-methylpyridine-3-boronicacid,alphachiron 13000b651,boronicacid, b-2-chloro-5-methyl-3-pyridinyl,2-chloro-5-methyl-3-pyridineboronic acid,pubchem11116,acmc-209ra8,abbypharma ap-16-5787
2763126
(2-chloro-5-methylpyridin-3-yl)boronic acid
B(C1=CC(=CN=C1Cl)C)(O)O
913835-86-6
C6H7BClNO2
171.39
ZMHWEBDZKPQNHG-UHFFFAOYSA-N
2-chloro-5-methylpyridine-3-boronic acid,2-chloro-5-methylpyridin-3-yl boronic acid,2-chloro-5-picoline-3-boronic acid,2-chloro-5-methylpyridine-3-boronicacid,alphachiron 13000b651,boronicacid, b-2-chloro-5-methyl-3-pyridinyl,2-chloro-5-methyl-3-pyridineboronic acid,pubchem11116,acmc-209ra8,abbypharma ap-16-5787
2763126
(2-chloro-5-methylpyridin-3-yl)boronic acid
B(C1=CC(=CN=C1Cl)C)(O)O
Packaging Glass Bottle
Quantity 1g
Melting Point 115.5°C to 119.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 171.39
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methylpyridine-3-boronic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning