Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-methylpyrimidine, 97%

Click to view available options
Quantity:
1g
5g
25g
22536-61-4
C5H5ClN2
128.559
MFCD09260903
APRMCBSTMFKLEI-UHFFFAOYSA-N
pyrimidine, 2-chloro-5-methyl,2-chloro-5-methyl-pyrimidine,pubchem11095,2-chloro-5-methyl pyrimidine,ksc497i7j
581719
2-chloro-5-methylpyrimidine
CC1=CN=C(N=C1)Cl
22536-61-4
C5H5ClN2
128.559
MFCD09260903
APRMCBSTMFKLEI-UHFFFAOYSA-N
pyrimidine, 2-chloro-5-methyl,2-chloro-5-methyl-pyrimidine,pubchem11095,2-chloro-5-methyl pyrimidine,ksc497i7j
581719
2-chloro-5-methylpyrimidine
CC1=CN=C(N=C1)Cl
Applications
Used in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 5g
Melting Point 89°C to 92°C
Beilstein 108732
Formula Weight 128.56
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-methylpyrimidine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger