Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-n-decylpyrimidine, 99%

Click to view available options
Quantity:
1g
5g
170434-06-7
C14H23ClN2
254.802
MFCD02683077
LJRSECCCAXFYFU-UHFFFAOYSA-N
2-chloro-5-n-decylpyrimidine
22099083
2-chloro-5-decylpyrimidine
CCCCCCCCCCC1=CN=C(N=C1)Cl
170434-06-7
C14H23ClN2
254.802
MFCD02683077
LJRSECCCAXFYFU-UHFFFAOYSA-N
2-chloro-5-n-decylpyrimidine
22099083
2-chloro-5-decylpyrimidine
CCCCCCCCCCC1=CN=C(N=C1)Cl
Quantity 1g
Formula Weight 254.8
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-n-decylpyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning