Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-nitrobenzonitrile, 99%

Click to view available options
Quantity:
5g
25g
100g
16588-02-6
C7H3ClN2O2
182.563
MFCD00007292
ZGILLTVEEBNDOB-UHFFFAOYSA-N
benzonitrile, 2-chloro-5-nitro,2-chlor-5-nitrobenzonitril german,5-nitro-2-chlorobenzonitrile,chembl52706,2-chloro-5-nitro-benzonitrile,2-chloro-5-nitrobenzenecarbonitrile,4-chloro-3-cyano-1-nitrobenzene,2-chlor-5-nitrobenzonitril,pubchem4740,2-chloro-5-nitrobenzonitril
85504
2-chloro-5-nitrobenzonitrile
C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl
16588-02-6
C7H3ClN2O2
182.563
MFCD00007292
ZGILLTVEEBNDOB-UHFFFAOYSA-N
benzonitrile, 2-chloro-5-nitro,2-chlor-5-nitrobenzonitril german,5-nitro-2-chlorobenzonitrile,chembl52706,2-chloro-5-nitro-benzonitrile,2-chloro-5-nitrobenzenecarbonitrile,4-chloro-3-cyano-1-nitrobenzene,2-chlor-5-nitrobenzonitril,pubchem4740,2-chloro-5-nitrobenzonitril
85504
2-chloro-5-nitrobenzonitrile
C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl
Quantity 100g
Melting Point 104°C to 107°C
Beilstein 1818642
Formula Weight 182.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-nitrobenzonitrile

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DI3040000