Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-nitronicotinic acid, 95%

Click to view available options
Quantity:
1g
5g
42959-38-6
C6H3ClN2O4
202.55
MFCD06658402
WOFZRBCITDVRON-UHFFFAOYSA-N
2-chloro-5-nitronicotinic acid,2-chloro-5-nitropyridine-3-carboxylic acid,2-chloro-5-nitro-nicotinic acid,3-carboxy-2-chloro-5-nitropyridine,3-carboxy-2-chloro-5-nitro pyridine,2-chloro-5-nitronicotinicacid,pubchem9631,acmc-209js2,ksc235o0f,chloro-5-nitro nicotinic acid
12542664
C1=C(C(=NC=C1[N+](=O)[O-])Cl)C(=O)O
42959-38-6
C6H3ClN2O4
202.55
MFCD06658402
WOFZRBCITDVRON-UHFFFAOYSA-N
2-chloro-5-nitronicotinic acid,2-chloro-5-nitropyridine-3-carboxylic acid,2-chloro-5-nitro-nicotinic acid,3-carboxy-2-chloro-5-nitropyridine,3-carboxy-2-chloro-5-nitro pyridine,2-chloro-5-nitronicotinicacid,pubchem9631,acmc-209js2,ksc235o0f,chloro-5-nitro nicotinic acid
12542664
C1=C(C(=NC=C1[N+](=O)[O-])Cl)C(=O)O
Melting Point 140°C to 143°C
Quantity 5g
Color White to Yellow
Sensitivity Moisture Sensitive
Formula Weight 202.55
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-nitronicotinic acid

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning