Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-5-pyrimidinecarbonyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
110099-99-5
C5H2Cl2N2O
176.984
JUYQWJULYXCBAV-UHFFFAOYSA-N
2-chloro-5-pyrimidinecarbonyl chloride,5-pyrimidinecarbonylchloride, 2-chloro,acmc-20mcwr,2-chloropyrimidine-5-carbonylchloride,5-pyrimidinecarbonyl chloride,2-chloro,5-pyrimidinecarbonyl chloride, 2-chloro,5-pyrimidinecarbonyl chloride, 2-chloro-9ci
10631129
2-chloropyrimidine-5-carbonyl chloride
C1=C(C=NC(=N1)Cl)C(=O)Cl
110099-99-5
C5H2Cl2N2O
176.984
JUYQWJULYXCBAV-UHFFFAOYSA-N
2-chloro-5-pyrimidinecarbonyl chloride,5-pyrimidinecarbonylchloride, 2-chloro,acmc-20mcwr,2-chloropyrimidine-5-carbonylchloride,5-pyrimidinecarbonyl chloride,2-chloro,5-pyrimidinecarbonyl chloride, 2-chloro,5-pyrimidinecarbonyl chloride, 2-chloro-9ci
10631129
2-chloropyrimidine-5-carbonyl chloride
C1=C(C=NC(=N1)Cl)C(=O)Cl
Melting Point 50.0°C to 52.0°C
Quantity 5g
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 176.99
Physical Form Powder or Low Melting Solid
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-pyrimidinecarbonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger