Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-(trifluoromethoxy)benzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
1022922-16-2
C7H5BClF3O3
240.369
MFCD06656267
BAEHYECKXGVYKR-UHFFFAOYSA-N
2-chloro-5-trifluoromethoxy phenylboronic acid,2-chloro-5-trifluoromethoxy phenyl boronic acid,2-chloro-5-trifluoromethoxyphenylboronic acid,2-chloro-5-trifluoromethoxy phenyl boronic acid,boronic acid, b-2-chloro-5-trifluoromethoxy phenyl,pubchem23778,acmc-1c1ll,ksc500s0d,2-chloro-5-trifluoromethoxy phenylboronicacid,2-chloro-5-trifluoromethoxy benzeneboronic acid
2763238
[2-chloro-5-(trifluoromethoxy)phenyl]boronic acid
B(C1=C(C=CC(=C1)OC(F)(F)F)Cl)(O)O
1022922-16-2
C7H5BClF3O3
240.369
MFCD06656267
BAEHYECKXGVYKR-UHFFFAOYSA-N
2-chloro-5-trifluoromethoxy phenylboronic acid,2-chloro-5-trifluoromethoxy phenyl boronic acid,2-chloro-5-trifluoromethoxyphenylboronic acid,2-chloro-5-trifluoromethoxy phenyl boronic acid,boronic acid, b-2-chloro-5-trifluoromethoxy phenyl,pubchem23778,acmc-1c1ll,ksc500s0d,2-chloro-5-trifluoromethoxy phenylboronicacid,2-chloro-5-trifluoromethoxy benzeneboronic acid
2763238
[2-chloro-5-(trifluoromethoxy)phenyl]boronic acid
B(C1=C(C=CC(=C1)OC(F)(F)F)Cl)(O)O
Quantity 5g
Formula Weight 240.37
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-(trifluoromethoxy)benzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning