Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Chloro-5'-(trifluoromethyl)acetophenone, 97+%

Click to view available options
Quantity:
1g
5g
71648-45-8
C9H6ClF3O
222.591
MFCD00045092
YRGBMTWHOFQSDJ-UHFFFAOYSA-N
2'-chloro-5'-trifluoromethyl acetophenone,1-2-chloro-5-trifluoromethyl phenyl ethanone,1-2-chloro-5-trifluoromethyl phenyl ethan-1-one,2-chloro-5-trifluoromethyl acetophenone,acetophenone, 2-chloro-5-trifluoromethyl,3-acetyl-4-chlorobenzotrifluoride,ethanone, 1-2-chloro-5-trifluoromethyl phenyl,1-acetyl-2-chloro-5-trifluoromethyl benzene,pubchem4210
144502
1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl
71648-45-8
C9H6ClF3O
222.591
MFCD00045092
YRGBMTWHOFQSDJ-UHFFFAOYSA-N
2'-chloro-5'-trifluoromethyl acetophenone,1-2-chloro-5-trifluoromethyl phenyl ethanone,1-2-chloro-5-trifluoromethyl phenyl ethan-1-one,2-chloro-5-trifluoromethyl acetophenone,acetophenone, 2-chloro-5-trifluoromethyl,3-acetyl-4-chlorobenzotrifluoride,ethanone, 1-2-chloro-5-trifluoromethyl phenyl,1-acetyl-2-chloro-5-trifluoromethyl benzene,pubchem4210
144502
1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl
Applications
2′-Chloro-5′-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.11g/L @ 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Quantity 1g
Solubility Information Sparingly soluble in water 0.11g/L @ 25°C.
Formula Weight 222.6
Percent Purity ≥97%
Chemical Name or Material 2'-Chloro-5'-(trifluoromethyl)acetophenone

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures