Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)benzaldehyde, 97+%

Click to view available options
Quantity:
1g
5g
82386-89-8
C8H4ClF3O
208.564
MFCD00134430
OZZOJJJYKYKBNH-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl benzaldehyde,benzaldehyde, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl-benzaldehyde,pubchem2881,acmc-209pop,ksc495k9f,timtec-bb sbb003466,4-chloro-3-formylbenzotrifluoride
688181
2-chloro-5-(trifluoromethyl)benzaldehyde
C1=CC(=C(C=C1C(F)(F)F)C=O)Cl
82386-89-8
C8H4ClF3O
208.564
MFCD00134430
OZZOJJJYKYKBNH-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl benzaldehyde,benzaldehyde, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl-benzaldehyde,pubchem2881,acmc-209pop,ksc495k9f,timtec-bb sbb003466,4-chloro-3-formylbenzotrifluoride
688181
2-chloro-5-(trifluoromethyl)benzaldehyde
C1=CC(=C(C=C1C(F)(F)F)C=O)Cl
Flash Point 89°C (103°F)
Quantity 1g
Boiling Point 42°C to 44°C (1.5mmHg)
Formula Weight 208.57
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzaldehyde

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning