Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)benzyl alcohol, 97+%

Click to view available options
Quantity:
1g
5g
64372-62-9
C8H6ClF3O
210.58
MFCD01631469
VEKMDPQNERMQMO-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl benzyl alcohol,2-chloro-5-trifluoromethylbenzyl alcohol,2-chloro-5-trifluoromethyl phenyl methanol,2-chloro-5-trifluoromethylbenzylalcohol,2-chloro-5-trifluoromethyl benzylalcohol,4-chloro-3-hydroxymethyl benzotrifluoride,2-chloro-5-trifluoromethyl benzenemethanol,benzenemethanol, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl phenyl methan-1-ol
2778110
[2-chloro-5-(trifluoromethyl)phenyl]methanol
C1=CC(=C(C=C1C(F)(F)F)CO)Cl
64372-62-9
C8H6ClF3O
210.58
MFCD01631469
VEKMDPQNERMQMO-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl benzyl alcohol,2-chloro-5-trifluoromethylbenzyl alcohol,2-chloro-5-trifluoromethyl phenyl methanol,2-chloro-5-trifluoromethylbenzylalcohol,2-chloro-5-trifluoromethyl benzylalcohol,4-chloro-3-hydroxymethyl benzotrifluoride,2-chloro-5-trifluoromethyl benzenemethanol,benzenemethanol, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl phenyl methan-1-ol
2778110
[2-chloro-5-(trifluoromethyl)phenyl]methanol
C1=CC(=C(C=C1C(F)(F)F)CO)Cl
Quantity 5g
Melting Point 57°C to 60°C
Formula Weight 210.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures