Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)phenylacetonitrile, 97%

Click to view available options
Quantity:
1g
5g
22902-88-1
C9H5ClF3N
219.591
MFCD01631472
KTMVZYQQKJZSFK-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl phenylacetonitrile,2-chloro-5-trifluoromethylbenzylcyanide,2-chloro-5-trifluoromethylbenzyl cyanide,2-2-chloro-5-trifluoromethyl phenyl acetonitrile,benzeneacetonitrile,2-chloro-5-trifluoromethyl,2-2-chloro-5-trifluoromethyl phenyl ethanenitrile,acmc-20aicj,4-chloro-3-cyanomethyl benzotrifluoride,2-chloro-5-trifluoromethyl-phenyl-acetonitrile,2-chloro-5-trifluoromethyl phenyl acetonitrile
2778124
2-[2-chloro-5-(trifluoromethyl)phenyl]acetonitrile
C1=CC(=C(C=C1C(F)(F)F)CC#N)Cl
22902-88-1
C9H5ClF3N
219.591
MFCD01631472
KTMVZYQQKJZSFK-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl phenylacetonitrile,2-chloro-5-trifluoromethylbenzylcyanide,2-chloro-5-trifluoromethylbenzyl cyanide,2-2-chloro-5-trifluoromethyl phenyl acetonitrile,benzeneacetonitrile,2-chloro-5-trifluoromethyl,2-2-chloro-5-trifluoromethyl phenyl ethanenitrile,acmc-20aicj,4-chloro-3-cyanomethyl benzotrifluoride,2-chloro-5-trifluoromethyl-phenyl-acetonitrile,2-chloro-5-trifluoromethyl phenyl acetonitrile
2778124
2-[2-chloro-5-(trifluoromethyl)phenyl]acetonitrile
C1=CC(=C(C=C1C(F)(F)F)CC#N)Cl
Quantity 1g
Melting Point 30°C to 34°C
Boiling Point 140°C to 142°C (12mmHg)
UN Number UN3439
Beilstein 1960651
Formula Weight 219.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenylacetonitrile

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.