Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
505084-58-2
C7H3ClF3NO2
225.551
MFCD01862019
OIODRBHIESSYRO-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl pyridine-4-carboxylic acid,2-chloro-5-trifluoromethyl isonicotinic acid,2-chloro-5-trifluoromethyl-4-pyridinecarboxylic acid,4-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl isonicotinic acid,4-pyridinecarboxylicacid, 2-chloro-5-trifluoromethyl,acmc-209kng,ksc268s9f,2-chloro-5-trifluoromethylisonicotinic acid,2-chloro-5-trifluoromethyl isonicotinicacid
11264608
2-chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
C1=C(C(=CN=C1Cl)C(F)(F)F)C(=O)O
505084-58-2
C7H3ClF3NO2
225.551
MFCD01862019
OIODRBHIESSYRO-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl pyridine-4-carboxylic acid,2-chloro-5-trifluoromethyl isonicotinic acid,2-chloro-5-trifluoromethyl-4-pyridinecarboxylic acid,4-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl isonicotinic acid,4-pyridinecarboxylicacid, 2-chloro-5-trifluoromethyl,acmc-209kng,ksc268s9f,2-chloro-5-trifluoromethylisonicotinic acid,2-chloro-5-trifluoromethyl isonicotinicacid
11264608
2-chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
C1=C(C(=CN=C1Cl)C(F)(F)F)C(=O)O
Applications
It plays an important role for intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 5g
Formula Weight 225.55
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning