Alfa Aesar™ 2-Chloro-6-fluoro-3-methoxyaniline, 97%

Manufacturer: Alfa Aesar™ H3274206

Quantity 5g

 View more versions of this product

Catalog No. AAH3274206

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C7H7ClFNO
    Formula Weight 175.59
    MDL Number MFCD09832265
    UN Number UN2811
    Synonym 2-Chloro-6-fluoro-m-anisidine
    Chemical Name or Material 2-Chloro-6-fluoro-3-methoxyaniline

    Specifications

    Formula Weight 175.59
    Molecular Formula C7H7ClFNO
    MDL Number MFCD09832265
    UN Number UN2811
    Synonym 2-Chloro-6-fluoro-m-anisidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Melting Point 37° to 40°C
    Quantity 5g
    Chemical Name or Material 2-Chloro-6-fluoro-3-methoxyaniline
    Assay 97%

    Description

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit