Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Chloro-6'-fluoro-3'-methylacetophenone, 97+%

Click to view available options
Quantity:
1g
5g
261762-63-4
C9H8ClFO
186.61
MFCD01631368
DNESTVGNBQZBHP-UHFFFAOYSA-N
1-2-chloro-6-fluoro-3-methylphenyl ethanone,2'-chloro-6'-fluoro-3'-methylacetophenone,2-chloro-6-fluoro-3-methylacetophenone,1-2-chloro-6-fluoro-3-methylphenyl ethan-1-one,1-acetyl-2-chloro-6-fluoro-3-methylbenzene,pubchem7113,acmc-1cmd5,1-2-chloro-6-fluoro-3-methyl-phenyl-ethanone,ethanone,1-2-chloro-6-fluoro-3-methylphenyl,2'-chloro-6'-fluoro-3'-methyl-acetophenone
2773655
1-(2-chloro-6-fluoro-3-methylphenyl)ethanone
CC1=C(C(=C(C=C1)F)C(=O)C)Cl
261762-63-4
C9H8ClFO
186.61
MFCD01631368
DNESTVGNBQZBHP-UHFFFAOYSA-N
1-2-chloro-6-fluoro-3-methylphenyl ethanone,2'-chloro-6'-fluoro-3'-methylacetophenone,2-chloro-6-fluoro-3-methylacetophenone,1-2-chloro-6-fluoro-3-methylphenyl ethan-1-one,1-acetyl-2-chloro-6-fluoro-3-methylbenzene,pubchem7113,acmc-1cmd5,1-2-chloro-6-fluoro-3-methyl-phenyl-ethanone,ethanone,1-2-chloro-6-fluoro-3-methylphenyl,2'-chloro-6'-fluoro-3'-methyl-acetophenone
2773655
1-(2-chloro-6-fluoro-3-methylphenyl)ethanone
CC1=C(C(=C(C=C1)F)C(=O)C)Cl
Quantity 1g
Formula Weight 186.61
Percent Purity ≥97%
Chemical Name or Material 2'-Chloro-6'-fluoro-3'-methylacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning