Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-fluoro-3-methylbenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
104451-99-2
C8H6ClFO
172.583
MFCD01631360
CSYKEPRGLYWJCW-UHFFFAOYSA-N
2-chloro-6-fluoro-3-methyl-benzaldehyde,benzaldehyde,2-chloro-6-fluoro-3-methyl,acmc-20anyq,pubchem2882,2-chloro-6-fluoro-m-tolualdehyde,2-fluoro-5-methyl-6-chlorobenzaldehyde,benzaldehyde, 2-chloro-6-fluoro-3-methyl,2-chloranyl-6-fluoranyl-3-methyl-benzaldehyde,2-chloro-6-fluoro-3-methylbenzaldehyde
736334
2-chloro-6-fluoro-3-methylbenzaldehyde
CC1=C(C(=C(C=C1)F)C=O)Cl
104451-99-2
C8H6ClFO
172.583
MFCD01631360
CSYKEPRGLYWJCW-UHFFFAOYSA-N
2-chloro-6-fluoro-3-methyl-benzaldehyde,benzaldehyde,2-chloro-6-fluoro-3-methyl,acmc-20anyq,pubchem2882,2-chloro-6-fluoro-m-tolualdehyde,2-fluoro-5-methyl-6-chlorobenzaldehyde,benzaldehyde, 2-chloro-6-fluoro-3-methyl,2-chloranyl-6-fluoranyl-3-methyl-benzaldehyde,2-chloro-6-fluoro-3-methylbenzaldehyde
736334
2-chloro-6-fluoro-3-methylbenzaldehyde
CC1=C(C(=C(C=C1)F)C=O)Cl
Packaging Glass bottle
Melting Point 46°C to 50°C
Infrared Spectrum Authentic
Color White
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula F(Cl)C6H2(CH3)CHO
Formula Weight 172.59
Physical Form Crystalline Solid
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzaldehyde
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.