Alfa Aesar™ 2'-Chloro-6'-fluoroacetophenone, 97%

Manufacturer: Alfa Aesar™ B2406706

Quantity 5g

 View more versions of this product

Catalog No. AAB2406706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87327-69-3
    Molecular Formula C8H6ClFO
    Formula Weight 172.59
    MDL Number MFCD01631390
    Beilstein 5862530
    Chemical Name or Material 2'-Chloro-6'-fluoroacetophenone

    Specifications

    Formula Weight 172.59
    Molecular Formula C8H6ClFO
    MDL Number MFCD01631390
    CAS 87327-69-3
    Beilstein 5862530
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Quantity 5g
    Chemical Name or Material 2'-Chloro-6'-fluoroacetophenone
    Assay 97%

    Description

    Applications
    2 ft.-Chloro-6 ft.-fluoroacetophenone is used in medicine & organic intermediate.

    Solubility
    Sparingly soluble in water.

    Notes
    Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit