Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Chloro-6'-fluoroacetophenone, 97%

Click to view available options
Quantity:
1g
5g
25g
87327-69-3
C8H6ClFO
172.583
MFCD01631390
DNVGZKIRMBCQEQ-UHFFFAOYSA-N
2'-chloro-6'-fluoroacetophenone,1-2-chloro-6-fluorophenyl ethanone,2-chloro-6-fluoroacetophenone,1-2-chloro-6-fluorophenyl ethan-1-one,ethanone, 1-2-chloro-6-fluorophenyl,pubchem7110,1-acetyl-2-chloro-6-fluorobenzene,1-acetyl-6-chloro-2-fluorobenzene,1-2-chloro-6-fluoro-phenyl-ethanone,ethanone,1-2-chloro-6-fluorophenyl
2773578
1-(2-chloro-6-fluorophenyl)ethanone
CC(=O)C1=C(C=CC=C1Cl)F
87327-69-3
C8H6ClFO
172.583
MFCD01631390
DNVGZKIRMBCQEQ-UHFFFAOYSA-N
2'-chloro-6'-fluoroacetophenone,1-2-chloro-6-fluorophenyl ethanone,2-chloro-6-fluoroacetophenone,1-2-chloro-6-fluorophenyl ethan-1-one,ethanone, 1-2-chloro-6-fluorophenyl,pubchem7110,1-acetyl-2-chloro-6-fluorobenzene,1-acetyl-6-chloro-2-fluorobenzene,1-2-chloro-6-fluoro-phenyl-ethanone,ethanone,1-2-chloro-6-fluorophenyl
2773578
1-(2-chloro-6-fluorophenyl)ethanone
CC(=O)C1=C(C=CC=C1Cl)F
Applications
2′-Chloro-6′-fluoroacetophenone is used in medicine & organic intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Quantity 25g
Beilstein 5862530
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 2'-Chloro-6'-fluoroacetophenone

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning