Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-fluoroaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
363-51-9
C6H5ClFN
145.56
MFCD00040309
ZJLAWMDJTMMTQB-UHFFFAOYSA-N
benzenamine, 2-chloro-6-fluoro,2-chloro-6-fluoro-aniline,2-chloro-6-fluoro-phenylamine,6-chloro-2-fluorophenylamine,pubchem1528,2-fluoro-6-chloroaniline,acmc-209im1,2-chloro-6-fluoro aniline,2-chloro-6-fluorobenzenamine,2-chloro-6-fluorophenylamine
2734205
2-chloro-6-fluoroaniline
C1=CC(=C(C(=C1)Cl)N)F
363-51-9
C6H5ClFN
145.56
MFCD00040309
ZJLAWMDJTMMTQB-UHFFFAOYSA-N
benzenamine, 2-chloro-6-fluoro,2-chloro-6-fluoro-aniline,2-chloro-6-fluoro-phenylamine,6-chloro-2-fluorophenylamine,pubchem1528,2-fluoro-6-chloroaniline,acmc-209im1,2-chloro-6-fluoro aniline,2-chloro-6-fluorobenzenamine,2-chloro-6-fluorophenylamine
2734205
2-chloro-6-fluoroaniline
C1=CC(=C(C(=C1)Cl)N)F
Refractive Index 1.5480 to 1.554
Density 1.3160g/mL
Boiling Point 67°C to 69°C (14.0mmHg)
Specific Gravity 1.316
Color Yellow
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 145.56
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-chloro-6-fluoroaniline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-657-1