Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-6-fluoroanisole, 97+%

Click to view available options
Quantity:
1g
5g
53145-38-3
C7H6ClFO
160.572
MFCD01631573
QASFEHCRPLPGES-UHFFFAOYSA-N
2-chloro-6-fluoroanisole,1-chloro-3-fluoro-2-methoxy-benzene,pubchem20256,acmc-1awun,2-chloro-6-fluoro anisole,1-fluoro-2-methoxy-3-chlorobenzene,2-chloro-6-fluorophenol, methyl ether,1-chloro-3-fluoro-2-methoxy-benzene; 1-chloro-3-fluoro-2-methoxybenzene; 2-chloro-6-fluorophenyl methyl ether; benzene, 1-chloro-3-fluoro-2-methoxy
2773581
1-chloro-3-fluoro-2-methoxybenzene
COC1=C(C=CC=C1Cl)F
53145-38-3
C7H6ClFO
160.572
MFCD01631573
QASFEHCRPLPGES-UHFFFAOYSA-N
2-chloro-6-fluoroanisole,1-chloro-3-fluoro-2-methoxy-benzene,pubchem20256,acmc-1awun,2-chloro-6-fluoro anisole,1-fluoro-2-methoxy-3-chlorobenzene,2-chloro-6-fluorophenol, methyl ether,1-chloro-3-fluoro-2-methoxy-benzene; 1-chloro-3-fluoro-2-methoxybenzene; 2-chloro-6-fluorophenyl methyl ether; benzene, 1-chloro-3-fluoro-2-methoxy
2773581
1-chloro-3-fluoro-2-methoxybenzene
COC1=C(C=CC=C1Cl)F
Quantity 1g
Formula Weight 160.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-6-fluoroanisole

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning