Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-6-fluorobenzonitrile, 98%

Click to view available options
Quantity:
10g
50g
668-45-1
C7H3ClFN
155.556
MFCD00001780
XPTAYRHLHAFUOS-UHFFFAOYSA-N
2-fluoro-6-chlorobenzonitrile,benzonitrile, 2-chloro-6-fluoro,2-chloro-6-fluoro-benzonitrile,ncr bg ff,pubchem3423,ksc490q6t,acmc-1b036,timtec-bb sbb042501,2-chloro-6-fluorobenzenecarbonitrile,6-chloro-2-fluorobenzenecarbonitrile
69587
2-chloro-6-fluorobenzonitrile
C1=CC(=C(C(=C1)Cl)C#N)F
668-45-1
C7H3ClFN
155.556
MFCD00001780
XPTAYRHLHAFUOS-UHFFFAOYSA-N
2-fluoro-6-chlorobenzonitrile,benzonitrile, 2-chloro-6-fluoro,2-chloro-6-fluoro-benzonitrile,ncr bg ff,pubchem3423,ksc490q6t,acmc-1b036,timtec-bb sbb042501,2-chloro-6-fluorobenzenecarbonitrile,6-chloro-2-fluorobenzenecarbonitrile
69587
2-chloro-6-fluorobenzonitrile
C1=CC(=C(C(=C1)Cl)C#N)F
Odor Characteristic
Quantity 50g
Boiling Point 104°C to 105°C (11mmHg)
Melting Point 56°C to 60°C
UN Number UN3439
Beilstein 1940332
Formula Weight 155.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzonitrile

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.