Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorobenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
668-45-1
C7H3ClFN
155.556
MFCD00001780
XPTAYRHLHAFUOS-UHFFFAOYSA-N
2-fluoro-6-chlorobenzonitrile,benzonitrile, 2-chloro-6-fluoro,2-chloro-6-fluoro-benzonitrile,ncr bg ff,pubchem3423,ksc490q6t,acmc-1b036,timtec-bb sbb042501,2-chloro-6-fluorobenzenecarbonitrile,6-chloro-2-fluorobenzenecarbonitrile
69587
2-chloro-6-fluorobenzonitrile
C1=CC(=C(C(=C1)Cl)C#N)F
668-45-1
C7H3ClFN
155.556
MFCD00001780
XPTAYRHLHAFUOS-UHFFFAOYSA-N
2-fluoro-6-chlorobenzonitrile,benzonitrile, 2-chloro-6-fluoro,2-chloro-6-fluoro-benzonitrile,ncr bg ff,pubchem3423,ksc490q6t,acmc-1b036,timtec-bb sbb042501,2-chloro-6-fluorobenzenecarbonitrile,6-chloro-2-fluorobenzenecarbonitrile
69587
2-chloro-6-fluorobenzonitrile
C1=CC(=C(C(=C1)Cl)C#N)F
Quantity 50g
Packaging Glass bottle
Color Beige to Colorless
Infrared Spectrum Authentic
Melting Point 55°C to 59°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(F)CN
Formula Weight 155.56
Physical Form Crystals, Crystalline Powder and/or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloro-6-fluorobenzonitrile, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-571-2

TSCA : TSCA