Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-6-fluorobenzoyl chloride, 98%

Click to view available options
Quantity:
5g
25g
79455-63-3
C7H3Cl2FO
192.998
MFCD00178746
GFNAJZAKJGKJCS-UHFFFAOYSA-N
2-chloro-6-fluorobenzoylchloride,2-chloro-6-fluorobenzene-1-carbonyl chloride,2-chloro-6-fluoro-benzoyl chloride,6-chloro-2-fluorobenzoyl chloride,benzoyl chloride, 2-chloro-6-fluoro,acmc-1behg,2-fluoro-6-chlorobenzoyl chloride,attercop-chm at108371,2-chloro-6-fluoro benzoyl chloride,2-chloranyl-6-fluoranyl-benzoyl chloride
2736547
2-chloro-6-fluorobenzoyl chloride
C1=CC(=C(C(=C1)Cl)C(=O)Cl)F
79455-63-3
C7H3Cl2FO
192.998
MFCD00178746
GFNAJZAKJGKJCS-UHFFFAOYSA-N
2-chloro-6-fluorobenzoylchloride,2-chloro-6-fluorobenzene-1-carbonyl chloride,2-chloro-6-fluoro-benzoyl chloride,6-chloro-2-fluorobenzoyl chloride,benzoyl chloride, 2-chloro-6-fluoro,acmc-1behg,2-fluoro-6-chlorobenzoyl chloride,attercop-chm at108371,2-chloro-6-fluoro benzoyl chloride,2-chloranyl-6-fluoranyl-benzoyl chloride
2736547
2-chloro-6-fluorobenzoyl chloride
C1=CC(=C(C(=C1)Cl)C(=O)Cl)F
Quantity 25g
UN Number UN3265
Beilstein 4980867
Formula Weight 193.01
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.