Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorobenzyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
56456-50-9
C7H6ClFO
160.572
MFCD00004608
ZZFCUESFXBCRSC-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl alcohol,2-chloro-6-fluorophenyl methanol,2-chloro-6-fluorobenzylalcohol,2-fluoro-6-chlorobenzyl alcohol,unii-5k0sc480hm,benzenemethanol, 2-chloro-6-fluoro,6-fluoro-2-chlorobenzyl alcohol,2-chloro-6-fluorobenzylic alcohol,pubchem3427,acmc-1axh3
91868
(2-chloro-6-fluorophenyl)methanol
C1=CC(=C(C(=C1)Cl)CO)F
56456-50-9
C7H6ClFO
160.572
MFCD00004608
ZZFCUESFXBCRSC-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl alcohol,2-chloro-6-fluorophenyl methanol,2-chloro-6-fluorobenzylalcohol,2-fluoro-6-chlorobenzyl alcohol,unii-5k0sc480hm,benzenemethanol, 2-chloro-6-fluoro,6-fluoro-2-chlorobenzyl alcohol,2-chloro-6-fluorobenzylic alcohol,pubchem3427,acmc-1axh3
91868
(2-chloro-6-fluorophenyl)methanol
C1=CC(=C(C(=C1)Cl)CO)F
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 41°C to 44°C
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula ClC6H3(F)CH2OH
Formula Weight 160.57
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 2-Chloro-6-fluorobenzyl alcohol, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 260-192-9

TSCA : TSCA