Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-fluorobenzyl bromide, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
68220-26-8
C7H5BrClF
223.47
IGUVNNXFTDCASP-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl bromide,2-bromomethyl-1-chloro-3-fluorobenzene,2-chloro-6-fluorobenzylbromide,2-bromomethyl-1-chloro-3-fluoro-benzene,benzene, 2-bromomethyl-1-chloro-3-fluoro,2-bromomethyl-3-chloro-1-fluorobenzene,alpha-bromo-2-chloro-6-fluorotoluene,pubchem4894,acmc-1bdpq
2773625
2-(bromomethyl)-1-chloro-3-fluorobenzene
C1=CC(=C(C(=C1)Cl)CBr)F
68220-26-8
C7H5BrClF
223.47
IGUVNNXFTDCASP-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl bromide,2-bromomethyl-1-chloro-3-fluorobenzene,2-chloro-6-fluorobenzylbromide,2-bromomethyl-1-chloro-3-fluoro-benzene,benzene, 2-bromomethyl-1-chloro-3-fluoro,2-bromomethyl-3-chloro-1-fluorobenzene,alpha-bromo-2-chloro-6-fluorotoluene,pubchem4894,acmc-1bdpq
2773625
2-(bromomethyl)-1-chloro-3-fluorobenzene
C1=CC(=C(C(=C1)Cl)CBr)F
Specific Gravity 1.629
Refractive Index 1.569
Density 1.6290g/mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 5g
Flash Point >110°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula Cl(F)C6H3CH2Br
Formula Weight 223.47
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 2-chloro-6-fluorobenzyl bromide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger