Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorobenzyl bromide, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
68220-26-8
C7H5BrClF
223.469
MFCD00040126
IGUVNNXFTDCASP-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl bromide,2-bromomethyl-1-chloro-3-fluorobenzene,2-chloro-6-fluorobenzylbromide,2-bromomethyl-1-chloro-3-fluoro-benzene,benzene, 2-bromomethyl-1-chloro-3-fluoro,2-bromomethyl-3-chloro-1-fluorobenzene,alpha-bromo-2-chloro-6-fluorotoluene,pubchem4894,acmc-1bdpq
2773625
2-(bromomethyl)-1-chloro-3-fluorobenzene
C1=CC(=C(C(=C1)Cl)CBr)F
68220-26-8
C7H5BrClF
223.469
MFCD00040126
IGUVNNXFTDCASP-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl bromide,2-bromomethyl-1-chloro-3-fluorobenzene,2-chloro-6-fluorobenzylbromide,2-bromomethyl-1-chloro-3-fluoro-benzene,benzene, 2-bromomethyl-1-chloro-3-fluoro,2-bromomethyl-3-chloro-1-fluorobenzene,alpha-bromo-2-chloro-6-fluorotoluene,pubchem4894,acmc-1bdpq
2773625
2-(bromomethyl)-1-chloro-3-fluorobenzene
C1=CC(=C(C(=C1)Cl)CBr)F
Quantity 25g
Flash Point >110°C (230°F)
UN Number UN3265
Formula Weight 223.48
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.