Alfa Aesar™ 2-Chloro-6-fluorobenzyl chloride, 98%

Manufacturer: Alfa Aesar™ A1175814

Quantity 25g

 View more versions of this product

Catalog No. AAA1175814

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 55117-15-2
    Molecular Formula C7H5Cl2F
    Formula Weight 179.02
    MDL Number MFCD00000896
    UN Number UN3265
    Beilstein 386840
    Synonym alpha,2-Dichloro-6-fluorotoluene
    Chemical Name or Material 2-Chloro-6-fluorobenzyl chloride

    Specifications

    Formula Weight 179.02
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Beilstein 386840
    Health Hazard 1 Danger
    Molecular Formula C7H5Cl2F
    MDL Number MFCD00000896
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    TSCA TSCA
    CAS 55117-15-2
    UN Number UN3265
    Synonym alpha,2-Dichloro-6-fluorotoluene
    Flash Point 93°C (199°F)
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Boiling Point 208° to 210°C
    Chemical Name or Material 2-Chloro-6-fluorobenzyl chloride
    Assay 98%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit