Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorophenethylamine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
149488-93-7
C8H9ClFN
173.62
MFCD00052682
NNEMMAAHBKHXLP-UHFFFAOYSA-N
2-chloro-6-fluorophenethylamine,2-2-chloro-6-fluorophenyl ethanamine,2-2-chloro-6-fluorophenyl ethylamine,benzeneethanamine,2-chloro-6-fluoro,2-2-chloro-6-fluoro-phenyl-ethylamine,2-2-chloro-6-fluorophenyl ethan-1-amine,2-6-chloro-2-fluorophenyl ethylamine,acmc-1c8qt,benzeneethanamine, 2-chloro-6-fluoro
2734094
2-(2-chloro-6-fluorophenyl)ethanamine
C1=CC(=C(C(=C1)Cl)CCN)F
149488-93-7
C8H9ClFN
173.62
MFCD00052682
NNEMMAAHBKHXLP-UHFFFAOYSA-N
2-chloro-6-fluorophenethylamine,2-2-chloro-6-fluorophenyl ethanamine,2-2-chloro-6-fluorophenyl ethylamine,benzeneethanamine,2-chloro-6-fluoro,2-2-chloro-6-fluoro-phenyl-ethylamine,2-2-chloro-6-fluorophenyl ethan-1-amine,2-6-chloro-2-fluorophenyl ethylamine,acmc-1c8qt,benzeneethanamine, 2-chloro-6-fluoro
2734094
2-(2-chloro-6-fluorophenyl)ethanamine
C1=CC(=C(C(=C1)Cl)CCN)F
Quantity 1g
Infrared Spectrum Authentic
Color Colorless to Pink-Red
Packaging Glass bottle
Refractive Index 1.5285 to 1.5305
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 173.62
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorophenethylamine, 99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

TSCA : TSCA